1-Boc-4-hydroxypiperidine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate
MQ100
495484-25G
495484-5G
SIAL495484-25GEA
119
EUR
SIAL495484-25G
SIAL495484-5G
1-Boc-4-hydroxypiperidine, Sigma-Aldrich®
1-Boc-4-hydroxypiperidine
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol |
MDL Number:
MFCD01075174 CAS Number: 109384-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...