α-tri-Calcium phosphate (α-phase), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
50553-10G-F 50553-1G-F
SIAL50553-10G-FEA 776 EUR
SIAL50553-10G-F SIAL50553-1G-F
α-tri-Calcium phosphate (α-phase), Sigma-Aldrich®
α-tri-Calcium phosphate (α-phase)
Formula: Ca₃O₈P₂
CAS Number: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741