1,2-Benzisothiazol-3(2H)-one, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Benzisothiazol-3-one
M-Clarity™ quality level = MQ100
561487-25G 561487-5G
SIAL561487-25GEA 190 EUR
SIAL561487-25G SIAL561487-5G
1,2-Benzisothiazol-3(2H)-one, Sigma-Aldrich®
1,2-Benzisothiazol-3(2H)-one
Formula: C₇H₅NOS
MW: 151.19 g/mol
MDL Number: MFCD00044001
CAS Number: 2634-33-5
EINECS: 220-120-9