2-Chloro-5-formylpyridine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 6-Chloronicotinaldehyde, 2-Chloropyridine-5-carboxaldehyde, 6-Chloropyridine-3-carboxaldehyde

596175-1G 596175-5G
SIAL596175-1GEA 34.6 EUR
SIAL596175-1G SIAL596175-5G
2-Chloro-5-formylpyridine, Sigma-Aldrich®
2-Chloro-5-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
MDL Number: MFCD03095223
CAS Number: 23100-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR