2,3-Dimethylpiperazine, Sigma-Aldrich®

Supplier: Merck

696102-1G 696102-250MG
SIAL696102-1GEA 269 EUR
SIAL696102-1G SIAL696102-250MG
2,3-Dimethylpiperazine, Sigma-Aldrich®
2,3-Dimethylpiperazine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD07373397
CAS Number: 84468-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR