DL-α-Aminoadipic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: DL-2-Aminoadipic acid, (RS)-2-Aminohexanedioic acid, H-DL-Homoglu-OH, 2-Aminohexanedioic acid, H-DL-Aad-OH

MQ200

A0637-10G A0637-1G A0637-5G
SIALA0637-10GEA 511 EUR
SIALA0637-10G SIALA0637-1G SIALA0637-5G
DL-α-Aminoadipic acid, Sigma-Aldrich®
DL-α-Aminoadipic acid
Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
MDL Number: MFCD00063119
CAS Number: 542-32-5
EINECS: 208-809-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR