L-α-Aminoadipic acid, Sigma-Aldrich®

Supplier: Merck

A7275-100MG A7275-1G A7275-250MG A7275-5G
SIALA7275-100MGEA 67.2 EUR
SIALA7275-100MG SIALA7275-1G SIALA7275-250MG SIALA7275-5G
L-α-Aminoadipic acid, Sigma-Aldrich®
L-α-Aminoadipic acid
Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
MDL Number: MFCD00002636
CAS Number: 1118-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR