Piperidine-N-carbaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Formylpiperidine, N-Formylpiperidine

M-Clarity™ quality level = MQ100

F17407-100G F17407-500G
SIALF17407-100GEA 66.8 EUR
SIALF17407-100G SIALF17407-500G
Piperidine-N-carbaldehyde, Sigma-Aldrich®
Piperidine-N-carbaldehyde
Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR