3-Picolyl chloride hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride
P43602-25G P43602-5G
SIALP43602-25GEA 96 EUR
SIALP43602-25G SIALP43602-5G
3-Picolyl chloride hydrochloride, Sigma-Aldrich®
3-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III