Pyridine-2-carbaldehyde, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde

M-Clarity™ quality level = MQ200

P62003-25G P62003-100G P62003-500G
SIALP62003-25GEA 34.1 EUR
SIALP62003-25G SIALP62003-100G SIALP62003-500G
Pyridine-2-carbaldehyde, Sigma-Aldrich®
Pyridine-2-carbaldehyde
Formula: C₆H₅NO
MW: 107.11 g/mol
MDL Number: MFCD00006290
CAS Number: 1121-60-4
EINECS: 214-333-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR