TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: TAPS sodium salt, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulphonate
M-Clarity™ quality level = MQ200
T0647-100G T0647-25G T0647-500G
SIALT0647-100GEA 184 EUR
SIALT0647-100G SIALT0647-25G SIALT0647-500G
TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt), Sigma-Aldrich®
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)
Formula: C₇H₁₆NNaO₆S
MW: 265.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069903
CAS Number: 91000-53-2