TES sodium salt (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt), Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)ethanesulphonate, TES sodium salt
M-Clarity™ quality level = MQ200
T0772-100G T0772-25G T0772-500G
SIALT0772-100GEA 322 EUR
SIALT0772-100G SIALT0772-25G SIALT0772-500G
TES sodium salt (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt), Sigma-Aldrich®
TES sodium salt (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt)
Formula: C₆H₁₄NNaO₆S
MW: 251.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065482
CAS Number: 70331-82-7