TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-{[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amino}-1-propanesulphonic acid, N-Tris(hydroxymethyl)methyl-3-aminopropanesulphonic acid

T5130-100G T5130-1KG T5130-250G T5130-25G T5130-5KG
SIALT5130-100GEA 140 EUR
SIALT5130-100G SIALT5130-1KG SIALT5130-250G SIALT5130-25G SIALT5130-5KG
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid), Sigma-Aldrich®
TAPS (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula: C₇H₁₇NO₆S
MW: 243.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007538
CAS Number: 29915-38-6
EINECS: 249-954-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR