TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt), Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
TAPS sodium salt, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulphonate
T5441-100G
T5441-500G
SIALT5441-100GEA
385
EUR
SIALT5441-100G
SIALT5441-500G
TAPS sodium salt (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt), Sigma-Aldrich®
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)
Formula:
C₇H₁₆NNaO₆S MW: 265.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069903 CAS Number: 91000-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...