TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid
T6541-100G
T6541-25G
SIALT6541-100GEA
552
EUR
SIALT6541-100G
SIALT6541-25G
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Formula:
C₆H₁₅NO₆S MW: 229.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007532 CAS Number: 7365-44-8 EINECS: 230-906-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...