3,3,5-Trimethylcyclohexanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
T75752-1L T75752-250ML
SIALT75752-1LEA 114 EUR
SIALT75752-1L SIALT75752-250ML
3,3,5-Trimethylcyclohexanone, Sigma-Aldrich®
3,3,5-Trimethylcyclohexanone
Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 188.8 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019466
CAS Number: 873-94-9
EINECS: 212-855-9