3-Methyl-1,2-cyclohexanedione ≥98%
Supplier: Thermo Scientific
B22623.06 B22623.14
B22623.06EA 68.1 EUR
B22623.06 B22623.14
3-Methyl-1,2-cyclohexanedione ≥98%
3-Methyl-1,2-cyclohexanedione
Purity: 98+% CAS No.: 3008-43-3 Molecular Formula: C7H10O2 Formula Weight: 126.16 Melting Point: 57-59° Boiling Point: 69-72°/1mm MDL No.: MFCD00209518
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 69…72 °C (1 mmHg)
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00209518
CAS Number: 3008-43-3
EINECS: 221-122-2