1,5-Diazobicyclo[4.3.0]-5-nonene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DBN, 2,3,4,6,7,8-Hexahydropyrrolo[1,2-a]pyrimidine, 1,5-Diazabicyclo[4.3.0]-5-nonene
M-Clarity™ quality level = MQ200
33471-100ML 33471-25ML 33471-500ML
SIAL33471-100MLEA 290 EUR
SIAL33471-100ML SIAL33471-25ML SIAL33471-500ML
1,5-Diazobicyclo[4.3.0]-5-nonene, Sigma-Aldrich®
1,5-Diazobicyclo[4.3.0]-5-nonene
Formula: C₇H₁₂N₂
MW: 124.19 g/mol
Boiling Pt: 110 °C (20 hPa)
Density: 1 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00005554
CAS Number: 3001-72-7
EINECS: 221-087-3
UN: 3267
ADR: 8,II