di-Sodium tetraborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Borax, Sodium tetraborate
M-Clarity™ quality level = MQ200
71997-100G 71997-1KG 71997-500G
SIAL71997-100GEA 107 EUR
SIAL71997-100G SIAL71997-1KG SIAL71997-500G
di-Sodium tetraborate, Sigma-Aldrich®
di-Sodium tetraborate
Formula: Na₂B₄O₇
MW: 201.22 g/mol
Boiling Pt: 1575 °C (1013 hPa)
Melting Pt: 742 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733