Potassium cyanide ≥96.0% ACS
Supplier: Thermo Fisher Scientific
Danger
12136.18 12136.30 12136.A1
12136.18EA 26.3 EUR
12136.18 12136.30 12136.A1
Potassium cyanide ≥96.0% ACS
Potassium cyanide
Formula: KCN
MW: 65.12 g/mol
Boiling Pt: 1625 °C
Melting Pt: 634
Density: 1.52
Storage Temperature: Ambient
MDL Number: MFCD00011397
CAS Number: 151-50-8
EINECS: 205-792-3
UN: 1680
ADR: 6.1 I
Merck Index: 14,7626