Lithium carbonate 99%, pure
Supplier: Thermo Scientific
Warning
197785000 197780025 197780100
ACRO197785000EA 159 EUR
ACRO197785000 ACRO197780025 ACRO197780100
Lithium carbonate 99%, pure
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2,11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration excess HCl ≥98.5 %
Loss on ignition ≤0.7 % (500°C, 30 min)