Potassium hydrogen carbonate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassium bicarbonate

60339-1KG 60339-500G
SIAL60339-1KGEA 103 EUR
SIAL60339-1KG SIAL60339-500G
Potassium hydrogen carbonate, Sigma-Aldrich®
Potassium hydrogen carbonate
Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR