Sodium hydrogen carbonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
71631-1KG 71631-25KG 71631-5KG
SIAL71631-1KGEA 40.5 EUR
SIAL71631-1KG SIAL71631-25KG SIAL71631-5KG
Sodium hydrogen carbonate, Sigma-Aldrich®
Sodium hydrogen carbonate
Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM