Sodium chloride ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Warning
|
Synonyms:
Saline, NaCl, Sea salt, Halite
12314.36
12314.A3
12314.A9
12314.36EA
50.1
EUR
12314.36
12314.A3
12314.A9
Sodium chloride ≥99.0% ACS
Sodium chloride
Purity: 99.0% min
CAS No.: 7647-14-5
Molecular Formula: NaCl
Formula Weight: 58.44
Physical Form: Crystalline
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Solubility: Soluble in water, glycerol. Very slightly soluble in alcohol
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 °C Melting Pt: 801 Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |