Sodium chloride, high purity
Supplier: VWR Chemicals
Synonyms: Saline, NaCl, Sea salt, Halite
X190-1KGEA 185 EUR
X190-1KG X190-5KG
Sodium chloride, high purity
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599