Sodium chloride, Sigma-Aldrich®
Supplier: Merck
71376-1KG 71376-5KG
SIAL71376-1KGEA 77.8 EUR
SIAL71376-1KG SIAL71376-5KG
Sodium chloride, Sigma-Aldrich®
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599