Potassium iodide, Sigma-Aldrich®
Supplier: Merck
60399-100G-F 60399-500G-F
SIAL60399-100G-FEA 157 EUR
SIAL60399-100G-F SIAL60399-500G-F
Potassium iodide, Sigma-Aldrich®
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727