Sodium periodate
Supplier: Hach
HACH2107769EA
45.2
EUR
HACH2107769
Sodium periodate
Sodium periodate
Formula:
NaIO₄ MW: 213,89 g/mol Boiling Pt: 300 °C (1013 hPa) Melting Pt: 175 °C Density: 3,87 g/cm³ (20 °C) |
MDL Number:
MFCD00003534 CAS Number: 7790-28-5 UN: 1479 ADR: 5.1,II Merck Index: 13,08714 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...