Periodic acid 99.0-101.0% ACS
Supplier: Thermo Scientific
30472.18EA 142 EUR
30472.18
Periodic acid 99.0-101.0% ACS
Periodic acid
Formula: H₅IO₆
MW: 227,94 g/mol
Melting Pt: 126…130 °C
Density: 1,3875 g/cm³ (20 °C)
MDL Number: MFCD00011344
CAS Number: 10450-60-9
UN: 3085
ADR: 5.1,II
Merck Index: 14,7171
https://si.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg