Potassium permanganate
Supplier: Roth Carl
Danger
Synonyms: Permanganic acid potassium salt
8004.1 8004.2 8004.3
ROTH8004.1EA 30.6 EUR
ROTH8004.1 ROTH8004.2 ROTH8004.3
Potassium permanganate
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824