Sodium azide ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Sodium triazide
14314.09
14314.22
14314.36
14314.09EA
43.6
EUR
14314.09
14314.22
14314.36
Sodium azide ≥99%
Sodium azide
Purity: 99%
CAS No.: 26628-22-8
Molecular Formula: N3Na
Formula Weight: 65.01
Physical Form: Powder or Granular
Density: 1.846
Appearance: White
Odour: Odourless
pH: 10
Melting Point: ca 275° dec
Solubility: Soluble in water and liquid ammonia. Slightly soluble in alcohol
Merck Reference: 14,8581
UN No.: 1687
MDL No.: MFCD00003536
In airbag inflation, as a preservative in diagnostic medicinals
Formula:
NaN₃ MW: 65.01 g/mol Melting Pt: ∼ 275 °C (dec.) Density: 1.846 Storage Temperature: Ambient |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 EINECS: 247-852-1 UN: 1687 ADR: 6.1,II Merck Index: 14,8581 |