Sodium azide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium triazide
71290-100G 71290-500G
SIAL71290-100GEA 51.8 EUR
SIAL71290-100G SIAL71290-500G
Sodium azide, Sigma-Aldrich®
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
Melting Pt: 275
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1 II
Merck Index: 13,08653