Potassium nitrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Nitrate of Potash
M-Clarity™ quality level = MQ100
204110-50G 204110-250G
SIAL204110-50GEA 203 EUR
SIAL204110-50G SIAL204110-250G
Potassium nitrate, Sigma-Aldrich®
Potassium nitrate
Formula: KNO₃
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 13,07733