Sodium nitrate, Sigma-Aldrich®
Supplier: Merck
Warning
71755-1KG 71755-250G
SIAL71755-1KGEA 174 EUR
SIAL71755-1KG SIAL71755-250G
Sodium nitrate, Sigma-Aldrich®
Sodium nitrate
Formula: NaNO₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720