Rubidium nitrate 99.8%, pure
Supplier: Thermo Fisher Scientific
Danger
300070100 300070500
ACRO300070100EA 59.2 EUR
ACRO300070100 ACRO300070500
Rubidium nitrate 99.8%, pure
Rubidium nitrate
Formula: NO₃Rb
MW: 147.47 g/mol
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3.11 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1 II
Order Now
Specification Test Results
Appearance White Crystals or needles
Total trace metal impurities ≤0.2 %