Water, Molecular Biology Grade
Supplier: PanReac AppliChem
Synonyms: Hydrogen oxide
A7398.0500 A7398.1000 A7398.5000
A7398.0500EA 29 EUR
A7398.0500 A7398.1000 A7398.5000
Water, Molecular Biology Grade
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
Order Now
Specification Test Results
DNases/RNases/Proteases not detectable