Water cell biology, sterile ultrafiltered, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hydrogen oxide

95289-100ML 95289-1L
SIAL95289-100MLEA 23.4 EUR
SIAL95289-100ML SIAL95289-1L
Water cell biology, sterile ultrafiltered, Sigma-Aldrich®
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR