Counting chambers, Imp. Neubauer
Supplier: MARIENFELD
0640010 0640011 0640031
631-0696EA 35.7 EUR
631-0696 MARI0640011 MARI0640031
Counting chambers, Imp. Neubauer
Slides Counting Chambers
These counting chambers are designed for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
Certifikace: IVD directive 98/79/EC.
Delivery information: Supplied with two cover glasses.