Replacement parts for screw joints, BOLA
Supplier: Bohlender
D502-05 D502-02 D501-01 D501-04 D502-04 D502-09 D502-08 D502-07 D502-06 D502-03 D502-01 D501-07
BOHLD502-05EA 5.1 EUR
BOHLD502-05 BOHLD502-02 BOHLD501-01 BOHLD501-04 BOHLD502-04 BOHLD502-09 BOHLD502-08 BOHLD502-07 BOHLD502-06 BOHLD502-03 BOHLD502-01 BOHLD501-07
Replacement parts for screw joints, BOLA
Fittings and Connectors
Made of glass-fibre reinforced PTFE.
  • Temperature resistance: from –200 to +250 °C
  • Very good chemical resistance
For fittings, valves and stopcocks.