Potassium cyanate 97%, pure
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cyanic acid potassium salt
201110010 201115000 201111000
ACRO201110010EA 91.3 EUR
ACRO201110010 ACRO201115000 ACRO201111000
Potassium cyanate 97%, pure
Potassium cyanate
Formula: CKNO
MW: 81.12 g/mol
Melting Pt: 315 °C
Storage Temperature: Ambient
MDL Number: MFCD00011411
CAS Number: 590-28-3
EINECS: 209-676-3
Merck Index: 12,07788
Order Now
Specification Test Results
Appearance White Crystalline powder and/or chunks
Titration Argentometric ≥96.0 %
Potassium carbonate ≤3 %