Tetrasodium diphosphate ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate
A17546.0B A17546.30
A17546.0BEA 67.1 EUR
A17546.0B A17546.30
Tetrasodium diphosphate ≥98%
tetra-Sodium diphosphate
Purity: 98% CAS No.: 7722-88-5 Molecular Formula: Na4O7P2 Formula Weight: 265.90 Melting Point: 988° Merck Reference: 14,9240 MDL No.: MFCD00003513
Formula: Na₄O₇P₂
MW: 265.9 g/mol
Melting Pt: 988 °C
Density: 2.534
Storage Temperature: Ambient
MDL Number: MFCD00003513
CAS Number: 7722-88-5
EINECS: 231-767-1
Merck Index: 14,9240