Disodium sulphide 90+%, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Sodium sulphide
310180100 310180500 310182500
ACRO310180100EA 135 EUR
ACRO310180100 ACRO310180500 ACRO310182500
Disodium sulphide 90+%, pure
Disodium sulphide
Formula: Na₂S
MW: 78.05 g/mol
Boiling Pt: 174
Storage Temperature: Refrigerator
MDL Number: MFCD00003498
CAS Number: 1313-82-2
EINECS: 215-211-5
UN: 1385
ADR: 4.2 II
REACH: 01-2119513694-38
Merck Index: 12,08830
Order Now
Specification Test Results
Appearance Yellow to beige Powder and/or lumps
Titration Iodimetric ≥90.0 %