Sodium peroxodisulphate, Sigma-Aldrich®
Supplier: Merck
Danger
71889-1KG 71889-5KG
SIAL71889-1KGEA 421 EUR
SIAL71889-1KG SIAL71889-5KG
Sodium peroxodisulphate, Sigma-Aldrich®
Sodium peroxodisulphate
Formula: Na₂S₂O₈
Melting Pt: 100 °C
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1,III
Merck Index: 13,08729