Sodium sulphite, Sigma-Aldrich®
Supplier: Merck
71988-1KG 71988-250G
SIAL71988-1KGEA 179 EUR
SIAL71988-1KG SIAL71988-250G
Sodium sulphite, Sigma-Aldrich®
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757