Lithium sulphate, Sigma-Aldrich®

Supplier: Merck
Warning

62613-1KG 62613-250G
SIAL62613-1KGEA 292 EUR
SIAL62613-1KG SIAL62613-250G
Lithium sulphate, Sigma-Aldrich®
Lithium sulphate
Formula: Li₂O₄S
MW: 109.95 g/mol
Boiling Pt: 1377 °C (1013 hPa)
Melting Pt: 859 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS Number: 10377-48-7
EINECS: 233-820-4
Merck Index: 12,05569

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR