Membrane disc filters, GN-4 Metricel® MCE
Supplier: Cytiva (Formerly Pall Lab)
516-7727EA 0 EUR
516-7727 516-9017 514-0464
Membrane disc filters, GN-4 Metricel® MCE
Filters Membrane Filters
Hydrophilic mixed cellulose esters GN-4 Metricel® membrane for air monitoring applications. Dissolves completely using standard digestion procedures and meets NIOSH requirements for airborne metals and asbestos monitoring.
  • Cleans completely, possesses low artifacts, and offers minimal interference in fibre counting