Sodium selenite, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
71950-100G 71950-500G
SIAL71950-100GEA 78.9 EUR
SIAL71950-100G SIAL71950-500G
Sodium selenite, Sigma-Aldrich®
Sodium selenite
Formula: Na₂SeO₃
Melting Pt: 710 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003489
CAS Number: 10102-18-8
EINECS: 233-267-9
UN: 2630
ADR: 6.1,I
Merck Index: 12,08822