Sodium telluride ≥99.9% (metals basis) -60 mesh
Supplier: Thermo Fisher Scientific
|
Danger
|
41777.02
41777.04
41777.09
41777.02EA
0
EUR
41777.02
41777.04
41777.09
Sodium telluride ≥99.9% (metals basis) -60 mesh
Sodium telluride
Formula:
Na₂Te MW: 173.58 g/mol Melting Pt: 953 °C Density: 2.90 Storage Temperature: Ambient |
CAS Number:
12034-41-2 EINECS: 234-806-0 UN: 3134 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...