Sodium metavanadate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium vanadium trioxide
72060-100G 72060-25G 72060-500G
SIAL72060-100GEA 91.8 EUR
SIAL72060-100G SIAL72060-25G SIAL72060-500G
Sodium metavanadate, Sigma-Aldrich®
Sodium metavanadate
Formula: NaO₃V
MW: 121.93 g/mol
Melting Pt: 630
Density: 5.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011125
CAS Number: 13718-26-8
EINECS: 237-272-7
UN: 3285
ADR: 6.1 III
Merck Index: 12,08849