Calcium chloride, anhydrous ≥93%, porous
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
33327.36 33327.A3
33327.36EA 53.5 EUR
33327.36 33327.A3
Calcium chloride, anhydrous ≥93%, porous
Calcium chloride
Purity: 93% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Physical Form: -4 Mesh Sensitivity: Hygroscopic Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: >1600 °C
Melting Pt: 782 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,1659